missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dimethylthiazole 99.0+%, TCI America™

Catalog No. D12205ML
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D12205ML 5 mL
D122025ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D12205ML Supplier TCI America Supplier No. D12205ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 541-58-2
Molecular Formula C5H7NS
Molecular Weight (g/mol) 113.18
MDL Number MFCD00014509
InChI Key OBSLLHNATPQFMJ-UHFFFAOYSA-N
Synonym 2,4-dimethylthiazole, thiazole, 2,4-dimethyl, 2,4-methylthiazole, 2,4-dimethyl thiazole, unii-lht97038oa, 2,4-dimethyl-thiazole, dimethylthiazol, zlchem 533, 2,4 dimethylthiazole, 2.4-dimethylthiazole
PubChem CID 10934
IUPAC Name 2,4-dimethyl-1,3-thiazole
SMILES CC1=NC(C)=CS1

Specifications

Color Yellow
Boiling Point 146°C
Quantity 5 mL
UN Number 1993
Formula Weight 113.18
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,4-Dimethylthiazole
RTECSNumber XJ4375000
TSCA Yes