missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dimethylthiazole, 99%

Catalog No. AAA1360414
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1360414 25 g
AAA1360422 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1360414 Supplier Thermo Scientific Chemicals Supplier No. A1360414
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 541-58-2 | C5H7NS | 113.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 541-58-2
Molecular Formula C5H7NS
Molecular Weight (g/mol) 113.18
MDL Number MFCD00014509
InChI Key OBSLLHNATPQFMJ-UHFFFAOYSA-N
Synonym 2,4-dimethylthiazole, thiazole, 2,4-dimethyl, 2,4-methylthiazole, 2,4-dimethyl thiazole, unii-lht97038oa, 2,4-dimethyl-thiazole, dimethylthiazol, zlchem 533, 2,4 dimethylthiazole, 2.4-dimethylthiazole
PubChem CID 10934
IUPAC Name 2,4-dimethyl-1,3-thiazole
SMILES CC1=NC(C)=CS1

Specifications

Density 1.057
Boiling Point 145°C to 147°C
Flash Point 42°C (107°F)
Odor Characteristic
Refractive Index 1.509
Quantity 25 g
UN Number UN1993
Beilstein 106414
Solubility Information Slightly soluble in water
Formula Weight 113.18
Percent Purity 99%
Chemical Name or Material 2,4-Dimethylthiazole
Show More Show Less
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 208-786-9
RTECSNumber XJ4375000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only