missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dimethylthiazole-5-carboxylic Acid 98.0+%, TCI America™

Catalog No. p-7128479
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D48131G 1 g
D48135G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D48131G Supplier TCI America Supplier No. D48131G
Only null left

Chemical Identifiers

CAS 53137-27-2
Molecular Formula C6H7NO2S
Molecular Weight (g/mol) 157.187
MDL Number MFCD00052943
InChI Key MQGBARXPCXAFRZ-UHFFFAOYSA-N
Synonym 2,4-dimethylthiazole-5-carboxylic acid, dimethyl-1,3-thiazole-5-carboxylic acid, 5-thiazolecarboxylic acid, 2,4-dimethyl, 2,4-dimethyl-5-thiazolecarboxylic acid, 2,4-dimethyl-thiazole-5-carboxylic acid, 2,4-dimethylthiazole-5-carboxylicacid, ksc269g8n, 2,4-dimethylthiazol-5-carboxylic acid, 2,4-dimethylthiazole-5carboxylic acid, 2 4-dimethylthiazole-5-carboxylic acid
PubChem CID 736488
IUPAC Name 2,4-dimethyl-1,3-thiazole-5-carboxylic acid
SMILES CC1=C(SC(=N1)C)C(=O)O

Specifications

Melting Point 234°C
Color White-Yellow
Quantity 1 g
Formula Weight 157.19
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,4-Dimethylthiazole-5-carboxylic Acid
EINECSNumber (5)-3403
TSCA No