missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4-Dimethylthiazole-5-carboxylic Acid 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 234°C |
| Color | White-Yellow |
| Quantity | 5 g |
| Formula Weight | 157.19 |
| Percent Purity | ≥98.0% (GC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 2,4-Dimethylthiazole-5-carboxylic Acid |
Safety and Handling
| EINECSNumber | (5)-3403 |
| TSCA | No |