missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dimethylquinoline 98.0+%, TCI America™

Catalog No. D12371G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D12371G 1 g
D12375G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D12371G Supplier TCI America Supplier No. D12371G

Chemical Identifiers

CAS 1198-37-4
Molecular Formula C11H11N
Molecular Weight (g/mol) 157.216
MDL Number MFCD00006760
InChI Key ZTNANFDSJRRZRJ-UHFFFAOYSA-N
PubChem CID 14536
IUPAC Name 2,4-dimethylquinoline
SMILES CC1=CC(=NC2=CC=CC=C12)C

Specifications

Color Yellow
Boiling Point 159°C
Quantity 1 g
Synonym 4-methylquinaldine, quinoline, 2,4-dimethyl, 2,4-dimethyl-quinoline, kryptidine, methylquinaldine, pubchem5875, quinoline,4-dimethyl, 2,4-dimethyl-quinolin, quinoline,2,4-dimethyl, acmc-1bq9j
Formula Weight 157.22
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material 2,4-Dimethylquinoline