missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dimethylquinoline, 95%

Catalog No. AAL08348MC
Change view
Click to view available options
Quantity:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL08348MC 100 mg
AAL08348ME 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL08348MC Supplier Thermo Scientific Chemicals Supplier No. L08348MC
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1198-37-4 | C11H11N | 157.216 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1198-37-4
Molecular Formula C11H11N
Molecular Weight (g/mol) 157.216
MDL Number MFCD00006760
InChI Key ZTNANFDSJRRZRJ-UHFFFAOYSA-N
PubChem CID 14536
IUPAC Name 2,4-dimethylquinoline
SMILES CC1=CC(=NC2=CC=CC=C12)C

Specifications

Density 1.06
Boiling Point 264°C to 265°C
Flash Point >110°C (230°F)
Refractive Index 1.605
Quantity 100 mg
Beilstein 114230
Synonym 4-methylquinaldine, quinoline, 2,4-dimethyl, 2,4-dimethyl-quinoline, kryptidine, methylquinaldine, pubchem5875, quinoline,4-dimethyl, 2,4-dimethyl-quinolin, quinoline,2,4-dimethyl, acmc-1bq9j
Formula Weight 157.21
Percent Purity 95%
Chemical Name or Material 2,4-Dimethylquinoline
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 214-832-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only