missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dimethylpentane 99.0+%, TCI America™

Catalog No. D07695ML
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D07695ML 5 mL
D076925ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D07695ML Supplier TCI America Supplier No. D07695ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 108-08-7
Molecular Formula C7H16
Molecular Weight (g/mol) 100.205
MDL Number MFCD00008945
InChI Key BZHMBWZPUJHVEE-UHFFFAOYSA-N
Synonym pentane, 2,4-dimethyl, unii-4jt8q9qohi, diisopropylmethane, 4jt8q9qohi, 2,4-dimethylpentane, standard for gc, dimethylpentane, heptan und isomere, 2,4-dimethyl-pentan, pentane,2,4-dimethyl, 2,4-dimethylpentane
PubChem CID 7907
IUPAC Name 2,4-dimethylpentane
SMILES CC(C)CC(C)C

Specifications

Color Colorless
Boiling Point 80°C
Quantity 5 mL
UN Number 1206
Formula Weight 100.21
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,4-Dimethylpentane
EINECSNumber (2)-0007
TSCA Yes