missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dimethylpentane, 98+%

Catalog No. AAL0357009
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0357009 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL0357009 Supplier Thermo Scientific Chemicals Supplier No. L0357009
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 108-08-7 | C7H16 | 100.205 g/mol

It is not only used for synthesizing WeiNiGuan, but also used in binder and coating industry. It is used for synthesizing resin fiber. It is also used as intermediates and adhesives in oil drop coagulation thickener.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is not only used for synthesizing WeiNiGuan, but also used in binder and coating industry. It is used for synthesizing resin fiber. It is also used as intermediates and adhesives in oil drop coagulation thickener.

Solubility
Soluble in acetone, benzene, chloroform, alcohol and ether.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 108-08-7
Molecular Formula C7H16
Molecular Weight (g/mol) 100.205
MDL Number MFCD00008945
InChI Key BZHMBWZPUJHVEE-UHFFFAOYSA-N
PubChem CID 7907
IUPAC Name 2,4-dimethylpentane
SMILES CC(C)CC(C)C

Specifications

Melting Point ∼-123°C
Density 0.67
Boiling Point 79°C to 80°C
Flash Point −12°C (10°F)
Refractive Index 1.382
Quantity 10 g
UN Number UN1206
Beilstein 1696855
Synonym pentane, 2,4-dimethyl, unii-4jt8q9qohi, diisopropylmethane, 4jt8q9qohi, 2,4-dimethylpentane, standard for gc, dimethylpentane, heptan und isomere, 2,4-dimethyl-pentan, pentane,2,4-dimethyl, 2,4-dimethylpentane
Solubility Information Soluble in acetone,benzene,chloroform,alcohol and ether.
Formula Weight 100.21
Percent Purity ≥98%
Chemical Name or Material 2,4-Dimethylpentane
Show More Show Less
Hazard Category H225-H304-H315-H319-H336
Hazard Statement GHS H Statement
H225-H304-H315-H336
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Causes skin irritation.
May cause drowsiness or dizziness.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P312-P331-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HEPTANES
EINECSNumber 203-548-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only