missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dimethylimidazole, 97%

Catalog No. p-7081802
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1194906 5 g
AAA1194914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1194906 Supplier Thermo Scientific Chemicals Supplier No. A1194906

CAS: 930-62-1 | C5H8N2 | 96.13 g/mol

It is employed as a curing agent of epoxy resins. It is also widely used in the bonding, coating, capsulation of epoxy resins, or used for composite materials. It is also employed as medical raw material or used in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a curing agent of epoxy resins. It is also widely used in the bonding, coating, capsulation of epoxy resins, or used for composite materials. It is also employed as medical raw material or used in organic synthesis.

Solubility
Soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed. Store in cool, dry conditions in well ventilated place.

Chemical Identifiers

CAS 930-62-1
Molecular Formula C5H8N2
Molecular Weight (g/mol) 96.13
MDL Number MFCD00022365
InChI Key LLPKQRMDOFYSGZ-UHFFFAOYSA-N
Synonym 2,4-dimethylimidazole, 2,4-dimethyl-1h-imidazole, 1h-imidazole, 2,4-dimethyl, 2,4-dimethyl-3h-imidazole, 2,4-dimethyl-1h-imidazol, imidazole, 2,4-dimethyl, 2,4-dimethyl, 2,5-dimethylimidazole, imidazole,4-dimethyl, pubchem18587
PubChem CID 70259
IUPAC Name 2,5-dimethyl-1H-imidazole
SMILES CC1=CN=C(C)N1

Specifications

Melting Point 84°C to 88°C
Boiling Point 266°C (733 mmHg)
Quantity 5 g
UN Number UN3263
Beilstein 1639
Solubility Information Soluble in water.
Formula Weight 96.13
Percent Purity 97%
Chemical Name or Material 2,4-Dimethylimidazole
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, BASIC, ORGANIC, N.O.S.
EINECSNumber 213-221-4
RTECSNumber NI4839712
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only