missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dimethylimidazole, 90%, Tech.

Catalog No. p-118046
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC210320050 5 g Glass bottle
AC210320250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC210320050 Supplier Thermo Scientific Chemicals Supplier No. 210320050
Only null left

CAS: 930-62-1 | C5H8N2 | 96.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 930-62-1
Molecular Formula C5H8N2
Molecular Weight (g/mol) 96.13
MDL Number MFCD00022365
InChI Key LLPKQRMDOFYSGZ-UHFFFAOYSA-N
Synonym 2,4-dimethylimidazole, 2,4-dimethyl-1h-imidazole, 1h-imidazole, 2,4-dimethyl, 2,4-dimethyl-3h-imidazole, 2,4-dimethyl-1h-imidazol, imidazole, 2,4-dimethyl, 2,4-dimethyl, 2,5-dimethylimidazole, imidazole,4-dimethyl, pubchem18587
PubChem CID 70259
IUPAC Name 2,5-dimethyl-1H-imidazole
SMILES CC1=CN=C(C)N1

Specifications

CAS Min % 88.0
CAS Max % 100.0
Melting Point 85.0°C to 87.0°C
Color Yellow to Orange
Boiling Point 266.0°C (733.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 88% min. (GC)
Packaging Glass bottle
Quantity 5 g
Beilstein 23, V,5, 172
Formula Weight 96.13
Percent Purity 90%
Grade Technical
Physical Form Viscous Liquid or Weak Mass
Chemical Name or Material 2, 4-Dimethylimidazole, tech., 90%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
EINECSNumber 213-221-4
TSCA TSCA

RUO – Research Use Only