missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dimethylhexane, Thermo Scientific™

Catalog No. AC469280010
Change view
Click to view available options
Quantity:
1 mL
5 mL
25 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC469280010 1 mL
AC469280050 5 mL
AC469280250 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC469280010 Supplier Thermo Scientific Chemicals Supplier No. 469280010
Only null left

Chemical Identifiers

CAS 589-43-5
Molecular Formula C8H18
Molecular Weight (g/mol) 114.23
MDL Number 00039929
InChI Key HDGQICNBXPAKLR-UHFFFAOYNA-N
IUPAC Name 2,4-dimethylhexane
SMILES CCC(C)CC(C)C

Specifications

Alpha Vector DIMETHYLHEXANE
Melting Point 108°C to 109°C
Color Colorless
Flash Point 10°C
Infrared Spectrum Conforms
Packaging Glass Bottle
Refractive Index 1.3940 to 1.3960 (20°C, 589 nm)
Quantity 1 mL
Solubility Information In water: (20°C) almost insoluble
Physical Form Liquid
Chemical Name or Material 2,4-Dimethylhexane
Show More Show Less
Hazard Category Flame
Hazard Statement Exclamation mark
EINECSNumber 209-649-6
Recommended Storage Normal conditions