missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dimethylhexane 98.0+%, TCI America™

Catalog No. D11105ML
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D11105ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D11105ML Supplier TCI America Supplier No. D11105ML
Only null left

Chemical Identifiers

CAS 589-43-5
Molecular Formula C8H18
Molecular Weight (g/mol) 114.23
MDL Number MFCD00039929
InChI Key HDGQICNBXPAKLR-UHFFFAOYNA-N
Synonym hexane, 2,4-dimethyl, alkanes, c7-10-iso, 2,4-dimethyl hexane, 2,4-dimethylhexane, acmc-1azfz, ksc269m2t, hexane, 2,4-dimethyl-8ci 9ci
PubChem CID 11511
IUPAC Name 2,4-dimethylhexane
SMILES CCC(C)CC(C)C

Specifications

Color Colorless
Boiling Point 109°C
Quantity 5 mL
UN Number 1262
Formula Weight 114.23
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,4-Dimethylhexane
EINECSNumber (2)-0008
TSCA No