missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dimethylheptane 98.0+%, TCI America™

Catalog No. d12041ml
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D12041ML 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D12041ML Supplier TCI America Supplier No. D12041ML
Only null left

Chemical Identifiers

CAS 2213-23-2
Molecular Formula C9H20
Molecular Weight (g/mol) 128.26
MDL Number MFCD00048738
InChI Key AUKVIBNBLXQNIZ-UHFFFAOYNA-N
PubChem CID 16656
IUPAC Name 2,4-dimethylheptane
SMILES CCCC(C)CC(C)C

Specifications

Color Colorless
Boiling Point 133°C
Quantity 1 mL
UN Number 1920
Formula Weight 128.26
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material 2,4-Dimethylheptane
EINECSNumber (2)-0009
TSCA No