missing translation for 'onlineSavingsMsg'
Learn More

2',4'-Dimethylacetophenone, 95%

Catalog No. aaa1299230
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
10 g
250 g
50 g
Catalog No. Quantity
AAA1299230 250 g
AAA1299209 10 g
AAA1299218 50 g
3 options

Catalog No. AAA1299230

Supplier: Thermo Scientific Chemicals A1299230

Only null left

CAS: 89-74-7 | C10H12O | 148.205 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 89-74-7
Molecular Formula C10H12O
Molecular Weight (g/mol) 148.205
MDL Number MFCD00003571
InChI Key HSDSKVWQTONQBJ-UHFFFAOYSA-N
Synonym 2',4'-dimethylacetophenone, 1-2,4-dimethylphenyl ethanone, 2,4-dimethylacetophenone, 4-acetyl-m-xylene, ethanone, 1-2,4-dimethylphenyl, acetyl-m-xylene, 1-2,4-dimethylphenyl ethan-1-one, acetophenone, 2',4'-dimethyl, unii-8k29me27ya, methyl 2,4-dimethylphenyl ketone
PubChem CID 6985
IUPAC Name 1-(2,4-dimethylphenyl)ethanone
SMILES CC1=CC(=C(C=C1)C(=O)C)C

Specifications

Density 0.999
Boiling Point 227°C to 228°C
Flash Point 100°C (212°F)
Odor Characteristic
Refractive Index 1.535
Quantity 250 g
Beilstein 956772
Formula Weight 148.21
Percent Purity 97%
Chemical Name or Material 2',4'-Dimethylacetophenone
Hazard Category H302+H332-H319
Precautionary Statement P261-P264b-P270-P271-P280i-P301+P312-P304+P340-P305+P351+P338-P312-P330-P501c
EINECSNumber 201-935-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only