missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4-dimethyl-3-furaldehyde, Thermo Scientific™
Specifications
Specifications
| Packaging | Amber Glass Bottle |
| Quantity | 1g |
| Formula Weight | 124.14 |
| Percent Purity | 95% |
| Chemical Name or Material | 2, 4-dimethyl-3-furaldehyde |