missing translation for 'onlineSavingsMsg'
Learn More

2,4-dimethyl-3-furaldehyde, Thermo Scientific™

Catalog No. CC65804CB
Change view
Click to view available options
Quantity:
250mg
1g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC65804CB 250mg Amber Glass Bottle
CC65804DA 1g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC65804CB Supplier Thermo Scientific Chemicals Supplier No. CC65804CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 75002-34-5
Molecular Formula C7H8O2
Molecular Weight (g/mol) 124.139
InChI Key MGSAYHDYMMHZQE-UHFFFAOYSA-N
Synonym 2,4-dimethyl-3-furaldehyde, 3-furancarbaldehyde,2,4-dimethyl, 3-furancarboxaldehyde,2,4-dimethyl, 3-furancarboxaldehyde, 2,4-dimethyl-9ci
PubChem CID 21702758
IUPAC Name 2,4-dimethylfuran-3-carbaldehyde
SMILES CC1=COC(=C1C=O)C

Specifications

Packaging Amber Glass Bottle
Quantity 250mg
Formula Weight 124.14
Percent Purity 95%
Chemical Name or Material 2, 4-dimethyl-3-furaldehyde