missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dimethyl-3-ethylpyrrole, 96%

Catalog No. p-117232
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC116210050 5 g
AC116210250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC116210050 Supplier Thermo Scientific Chemicals Supplier No. 116210050

CAS: 517-22-6 | C8H13N | 123.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 517-22-6
MDL Number MFCD00005222
InChI Key ZEBBLOXDLGIMEG-UHFFFAOYSA-N
Synonym 2,4-dimethyl-3-ethylpyrrole, kryptopyrrole, kryptopyrrol, 3-ethyl-2,4-dimethylpyrrole, cryptopyrrole, 2,4-dimethyl-3-ethyl-1h-pyrrole, 1h-pyrrole, 3-ethyl-2,4-dimethyl, pyrrole, 3-ethyl-2,4-dimethyl, acmc-209kvh, 5-20-05-00099 beilstein handbook reference
PubChem CID 10600
IUPAC Name 3-ethyl-2,4-dimethyl-1H-pyrrole
SMILES CCC1=C(NC=C1C)C

Specifications

CAS Min % 95.0
CAS Max % 100.0
Density 0.9130g/mL
Boiling Point 197°C (710.0 mmHg)
Flash Point 71°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Molecular Formula C8H13N
Refractive Index 1.4960 to 1.498
Quantity 5 g
Beilstein 20, I, 47
Specific Gravity 0.913
Molecular Weight (g/mol) 123.2
Formula Weight 123.2
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material 2, 4-Dimethyl-3-ethylpyrrole, 96%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 208-234-7
RTECSNumber UX9629500
TSCA TSCA
Recommended Storage Product darkens on standing

RUO – Research Use Only