missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dimethyl-1,3-oxazole-5-carboxylic acid, 97%

Catalog No. AC457350010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC457350010 1 g
AC457350050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC457350010 Supplier Thermo Scientific Chemicals Supplier No. 457350010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 2510-37-4 | C6H7NO3 | 141.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2510-37-4
Molecular Formula C6H7NO3
Molecular Weight (g/mol) 141.13
InChI Key JLSFKHJNJFXGAB-UHFFFAOYSA-N
Synonym 2,4-dimethyloxazole-5-carboxylic acid, 2,4-dimethyl-oxazole-5-carboxylic acid, dimethyl-1,3-oxazole-5-carboxylic acid, 2,4-dimethyl-1,3-oxazole-5-carboxylicacid, 2,4-dimethyl-5-oxazolecarboxylic acid, 5-oxazolecarboxylic acid, 2,4-dimethyl, pubchem17689, ukrorgsyn-bb bbv-156622, 2,4-dimethyl-oxazole-5-carboxylicacid, saltdata: free
PubChem CID 1132979
IUPAC Name 2,4-dimethyl-1,3-oxazole-5-carboxylic acid
SMILES CC1=C(OC(=N1)C)C(=O)O

Specifications

Melting Point 229.0°C to 231.0°C
Assay Percent Range 97%
Packaging Glass Bottle
Quantity 1 g
Formula Weight 141.13
Percent Purity 97%
Chemical Name or Material 2,4-Dimethyl-1,3-oxazole-5-carboxylic acid
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Harmful if inhaled.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

RUO – Research Use Only