missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4-Dimethoxypyrimidine 98.0+%, TCI America™
$68.97 - $203.17
Chemical Identifiers
| CAS | 3551-55-1 |
|---|---|
| Molecular Formula | C6H8N2O2 |
| Molecular Weight (g/mol) | 140.142 |
| MDL Number | MFCD00056028 |
| InChI Key | KEVRHVMWBKFGLO-UHFFFAOYSA-N |
| Synonym | pyrimidine, 2,4-dimethoxy, unii-980k6dh82b, dimethoxypyrimidine, zlchem 892, pubchem7104, acmc-1adzt, 2,4-dimethoxy-pyrimidine, ksc911k2l, 2,4-dimethoxypyrimidine, kevrhvmwbkfglo-uhfffaoysa |
| PubChem CID | 137976 |
| IUPAC Name | 2,4-dimethoxypyrimidine |
| SMILES | COC1=NC(=NC=C1)OC |
Chemical Identifiers
| 3551-55-1 | |
| 140.142 | |
| KEVRHVMWBKFGLO-UHFFFAOYSA-N | |
| 137976 | |
| COC1=NC(=NC=C1)OC |
| C6H8N2O2 | |
| MFCD00056028 | |
| pyrimidine, 2,4-dimethoxy, unii-980k6dh82b, dimethoxypyrimidine, zlchem 892, pubchem7104, acmc-1adzt, 2,4-dimethoxy-pyrimidine, ksc911k2l, 2,4-dimethoxypyrimidine, kevrhvmwbkfglo-uhfffaoysa | |
| 2,4-dimethoxypyrimidine |
Specifications
| 3551-55-1 | |
| 205°C | |
| MFCD00056028 | |
| pyrimidine, 2,4-dimethoxy, unii-980k6dh82b, dimethoxypyrimidine, zlchem 892, pubchem7104, acmc-1adzt, 2,4-dimethoxy-pyrimidine, ksc911k2l, 2,4-dimethoxypyrimidine, kevrhvmwbkfglo-uhfffaoysa | |
| COC1=NC(=NC=C1)OC | |
| 140.142 | |
| 140.14 | |
| Liquid |
| Colorless | |
| C6H8N2O2 | |
| 5 g | |
| KEVRHVMWBKFGLO-UHFFFAOYSA-N | |
| 2,4-dimethoxypyrimidine | |
| 137976 | |
| ≥98.0% (GC,T) | |
| 2,4-Dimethoxypyrimidine |
Safety and Handling
TSCA : No