missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dimethoxypyrimidine 98.0+%, TCI America™

Catalog No. D23275G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D23275G 5 g
D232725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D23275G Supplier TCI America Supplier No. D23275G

Chemical Identifiers

CAS 3551-55-1
Molecular Formula C6H8N2O2
Molecular Weight (g/mol) 140.142
MDL Number MFCD00056028
InChI Key KEVRHVMWBKFGLO-UHFFFAOYSA-N
Synonym pyrimidine, 2,4-dimethoxy, unii-980k6dh82b, dimethoxypyrimidine, zlchem 892, pubchem7104, acmc-1adzt, 2,4-dimethoxy-pyrimidine, ksc911k2l, 2,4-dimethoxypyrimidine, kevrhvmwbkfglo-uhfffaoysa
PubChem CID 137976
IUPAC Name 2,4-dimethoxypyrimidine
SMILES COC1=NC(=NC=C1)OC

Specifications

Color Colorless
Boiling Point 205°C
Quantity 5 g
Formula Weight 140.14
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2,4-Dimethoxypyrimidine