missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dimethoxypyrimidine, 98+%

CAS: 3551-55-1 | C6H8N2O2 | 140.142 g/mol

$50.84 - $170.19

Chemical Identifiers

CAS 3551-55-1
Molecular Formula C6H8N2O2
Molecular Weight (g/mol) 140.142
MDL Number MFCD00056028
InChI Key KEVRHVMWBKFGLO-UHFFFAOYSA-N
Synonym pyrimidine, 2,4-dimethoxy, unii-980k6dh82b, dimethoxypyrimidine, zlchem 892, pubchem7104, acmc-1adzt, 2,4-dimethoxy-pyrimidine, ksc911k2l, 2,4-dimethoxypyrimidine, kevrhvmwbkfglo-uhfffaoysa
PubChem CID 137976
IUPAC Name 2,4-dimethoxypyrimidine
SMILES COC1=NC(=NC=C1)OC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH2579903
View Documents
Thermo Scientific Chemicals
H2579903
1 g
Each for $50.84
Only null left
 
AAH2579906
View Documents
Thermo Scientific Chemicals
H2579906
5 g
Each for $170.19
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

3551-55-1
140.142
KEVRHVMWBKFGLO-UHFFFAOYSA-N
137976
COC1=NC(=NC=C1)OC
C6H8N2O2
MFCD00056028
pyrimidine, 2,4-dimethoxy, unii-980k6dh82b, dimethoxypyrimidine, zlchem 892, pubchem7104, acmc-1adzt, 2,4-dimethoxy-pyrimidine, ksc911k2l, 2,4-dimethoxypyrimidine, kevrhvmwbkfglo-uhfffaoysa
2,4-dimethoxypyrimidine

Specifications

3551-55-1
1.1501
93°C (199°F)
1.502
1 g
KEVRHVMWBKFGLO-UHFFFAOYSA-N
2,4-dimethoxypyrimidine
137976
≥98%
17°C
204°C to 205°C
C6H8N2O2
MFCD00056028
pyrimidine, 2,4-dimethoxy, unii-980k6dh82b, dimethoxypyrimidine, zlchem 892, pubchem7104, acmc-1adzt, 2,4-dimethoxy-pyrimidine, ksc911k2l, 2,4-dimethoxypyrimidine, kevrhvmwbkfglo-uhfffaoysa
COC1=NC(=NC=C1)OC
140.142
140.14
2,4-Dimethoxypyrimidine
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only