missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dimethoxypyrimidine, 98+%

Catalog No. AAH2579906
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2579906 5 g
AAH2579903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2579906 Supplier Thermo Scientific Chemicals Supplier No. H2579906

CAS: 3551-55-1 | C6H8N2O2 | 140.142 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3551-55-1
Molecular Formula C6H8N2O2
Molecular Weight (g/mol) 140.142
MDL Number MFCD00056028
InChI Key KEVRHVMWBKFGLO-UHFFFAOYSA-N
Synonym pyrimidine, 2,4-dimethoxy, unii-980k6dh82b, dimethoxypyrimidine, zlchem 892, pubchem7104, acmc-1adzt, 2,4-dimethoxy-pyrimidine, ksc911k2l, 2,4-dimethoxypyrimidine, kevrhvmwbkfglo-uhfffaoysa
PubChem CID 137976
IUPAC Name 2,4-dimethoxypyrimidine
SMILES COC1=NC(=NC=C1)OC

Specifications

Melting Point 17°C
Density 1.1501
Boiling Point 204°C to 205°C
Flash Point 93°C (199°F)
Refractive Index 1.502
Quantity 5 g
Formula Weight 140.14
Percent Purity ≥98%
Chemical Name or Material 2,4-Dimethoxypyrimidine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only