missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4-Dimethoxyphenylboronic Acid (contains varying amounts of Anhydride), TCI America™
$42.87 - $42.87
Chemical Identifiers
| CAS | 133730-34-4 |
|---|---|
| Molecular Formula | C8H11BO4 |
| MDL Number | MFCD01074590 |
| InChI Key | SQTUYFKNCCBFRR-UHFFFAOYSA-N |
| PubChem CID | 2734341 |
| IUPAC Name | (2,4-dimethoxyphenyl)boronic acid |
Chemical Identifiers
| 133730-34-4 | |
| MFCD01074590 | |
| 2734341 |
| C8H11BO4 | |
| SQTUYFKNCCBFRR-UHFFFAOYSA-N | |
| (2,4-dimethoxyphenyl)boronic acid |
Specifications
| 133730-34-4 | |
| Green-White | |
| MFCD01074590 | |
| 2,4-dimethoxybenzeneboronic acid, 2,4-dimethoxyphenyl boronic acid, 2,4-dimethoxyphenyl boranediol, boronic acid, 2,4-dimethoxyphenyl, 2,4-dimethoxybenzene boronic acid, boronic acid, b-2,4-dimethoxyphenyl, pubchem1823, acmc-209btd, ksc174i6j, 2,4-dimethoxyphenylboronicacid | |
| B(C1=C(C=C(C=C1)OC)OC)(O)O | |
| 181.982 | |
| 181.98 | |
| 2,4-Dimethoxyphenylboronic Acid (contains varying amounts of Anhydride) |
| 105°C | |
| C8H11BO4 | |
| 1 g | |
| SQTUYFKNCCBFRR-UHFFFAOYSA-N | |
| (2,4-dimethoxyphenyl)boronic acid | |
| 2734341 | |
| Crystalline Powder |
Safety and Handling
TSCA : No