missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dimethoxyphenylboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. D35211G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D35211G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D35211G Supplier TCI America Supplier No. D35211G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 133730-34-4
Molecular Formula C8H11BO4
MDL Number MFCD01074590
InChI Key SQTUYFKNCCBFRR-UHFFFAOYSA-N
PubChem CID 2734341
IUPAC Name (2,4-dimethoxyphenyl)boronic acid

Specifications

Melting Point 105°C
Color Green-White
Quantity 1 g
Synonym 2,4-dimethoxybenzeneboronic acid, 2,4-dimethoxyphenyl boronic acid, 2,4-dimethoxyphenyl boranediol, boronic acid, 2,4-dimethoxyphenyl, 2,4-dimethoxybenzene boronic acid, boronic acid, b-2,4-dimethoxyphenyl, pubchem1823, acmc-209btd, ksc174i6j, 2,4-dimethoxyphenylboronicacid
SMILES B(C1=C(C=C(C=C1)OC)OC)(O)O
Molecular Weight (g/mol) 181.982
Formula Weight 181.98
Physical Form Crystalline Powder
Chemical Name or Material 2,4-Dimethoxyphenylboronic Acid (contains varying amounts of Anhydride)