Learn More
2,4-Dimethoxyphenyl isocyanate, 97%
CAS: 84370-87-6 | C9H9NO3 | 179.175 g/mol
$44.66 - $129.59
Chemical Identifiers
| CAS | 84370-87-6 |
|---|---|
| Molecular Formula | C9H9NO3 |
| Molecular Weight (g/mol) | 179.175 |
| MDL Number | MFCD00002005 |
| InChI Key | WRAZHLRMDRTLOZ-UHFFFAOYSA-N |
| Synonym | 2,4-dimethoxyphenyl isocyanate, 2,4-dimethoxyphenylisocyanate, benzene, 1-isocyanato-2,4-dimethoxy, 2,4-dimethoxybenzenisocyanate, acmc-20aocl, 1-isocyanato-2,4-dimethoxy-benzene, #, benzene,1-isocyanato-2,4-dimethoxy, 1-isocyanato-2,4-bis methyloxy benzene |
| PubChem CID | 601637 |
| IUPAC Name | 1-isocyanato-2,4-dimethoxybenzene |
| SMILES | COC1=CC(=C(C=C1)N=C=O)OC |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAL0976203
|
Thermo Scientific Chemicals
L0976203 |
1 g |
Each for $44.66
|
|
|||||
|
AAL0976206
|
Thermo Scientific Chemicals
L0976206 |
5 g |
Each for $129.59
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 84370-87-6 | |
| 179.175 | |
| WRAZHLRMDRTLOZ-UHFFFAOYSA-N | |
| 601637 | |
| COC1=CC(=C(C=C1)N=C=O)OC |
| C9H9NO3 | |
| MFCD00002005 | |
| 2,4-dimethoxyphenyl isocyanate, 2,4-dimethoxyphenylisocyanate, benzene, 1-isocyanato-2,4-dimethoxy, 2,4-dimethoxybenzenisocyanate, acmc-20aocl, 1-isocyanato-2,4-dimethoxy-benzene, #, benzene,1-isocyanato-2,4-dimethoxy, 1-isocyanato-2,4-bis methyloxy benzene | |
| 1-isocyanato-2,4-dimethoxybenzene |
Specifications
| 84370-87-6 | |
| 139°C to 141°C (11 mmHg) | |
| C9H9NO3 | |
| MFCD00002005 | |
| UN2811 | |
| Moisture sensitive | |
| WRAZHLRMDRTLOZ-UHFFFAOYSA-N | |
| 1-isocyanato-2,4-dimethoxybenzene | |
| 601637 | |
| 97% |
| 31°C to 32°C | |
| >110°C (230°F) | |
| 1.5475 | |
| 1 g | |
| 1106649 | |
| 2,4-dimethoxyphenyl isocyanate, 2,4-dimethoxyphenylisocyanate, benzene, 1-isocyanato-2,4-dimethoxy, 2,4-dimethoxybenzenisocyanate, acmc-20aocl, 1-isocyanato-2,4-dimethoxy-benzene, #, benzene,1-isocyanato-2,4-dimethoxy, 1-isocyanato-2,4-bis methyloxy benzene | |
| COC1=CC(=C(C=C1)N=C=O)OC | |
| 179.175 | |
| 179.18 | |
| 2,4-Dimethoxyphenyl isocyanate |
Safety and Handling
GHS H Statement
H331-H334-H302-H315-H319-H335
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P272-P280-P285-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P333+P313-P342+P311-P362-P501c
H302+H312+H332-H315-H317-H319-H334-H335
DOTInformation : Hazard Class: 9; Packaging Group: III
EINECSNumber : 282-747-4
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only