missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dimethoxyphenyl isocyanate, 97%

Catalog No. AAL0976206
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0976206 5 g
AAL0976203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0976206 Supplier Thermo Scientific Chemicals Supplier No. L0976206
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 84370-87-6 | C9H9NO3 | 179.175 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 84370-87-6
Molecular Formula C9H9NO3
Molecular Weight (g/mol) 179.175
MDL Number MFCD00002005
InChI Key WRAZHLRMDRTLOZ-UHFFFAOYSA-N
Synonym 2,4-dimethoxyphenyl isocyanate, 2,4-dimethoxyphenylisocyanate, benzene, 1-isocyanato-2,4-dimethoxy, 2,4-dimethoxybenzenisocyanate, acmc-20aocl, 1-isocyanato-2,4-dimethoxy-benzene, #, benzene,1-isocyanato-2,4-dimethoxy, 1-isocyanato-2,4-bis methyloxy benzene
PubChem CID 601637
IUPAC Name 1-isocyanato-2,4-dimethoxybenzene
SMILES COC1=CC(=C(C=C1)N=C=O)OC

Specifications

Melting Point 31°C to 32°C
Boiling Point 139°C to 141°C (11 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.5475
Quantity 5 g
UN Number UN2811
Beilstein 1106649
Sensitivity Moisture sensitive
Formula Weight 179.18
Percent Purity 97%
Chemical Name or Material 2,4-Dimethoxyphenyl isocyanate
Show More Show Less
Hazard Category H302+H312+H332-H315-H317-H319-H334-H335
Hazard Statement GHS H Statement
H331-H334-H302-H315-H319-H335
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P272-P280-P285-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P333+P313-P342+P311-P362-P501c
DOTInformation Hazard Class: 9; Packaging Group: III
EINECSNumber 282-747-4
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only