missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dimethoxybenzyl alcohol, 99%, Thermo Scientific™

Chemical Identifiers

CAS 7314-44-5
Molecular Formula C9H12O3
Molecular Weight (g/mol) 168.19
MDL Number MFCD00004614
InChI Key RNKOUSCCPHSCFE-UHFFFAOYSA-N
Synonym 2,4-dimethoxybenzyl alcohol, 2,4-dimethoxyphenyl methanol, benzenemethanol, 2,4-dimethoxy, 2,4-dimethoxybenzylalcohol, 1-hydroxymethyl-2,4-dimethoxybenzene, 2,4-dimethoxyphenyl methan-1-ol, pubchem7454, acmc-209opp, 2,4-dimethoxy-benzylalcohol, 2,4-dimethoxybenzyl-alcohol
PubChem CID 81750
IUPAC Name (2,4-dimethoxyphenyl)methanol
SMILES COC1=CC(=C(C=C1)CO)OC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC170460250
SDS View Documents
Thermo Scientific Chemicals
170460250
25g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC170461000
SDS View Documents
Thermo Scientific Chemicals
170461000
100g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

7314-44-5
168.19
RNKOUSCCPHSCFE-UHFFFAOYSA-N
81750
COC1=CC(=C(C=C1)CO)OC
C9H12O3
MFCD00004614
2,4-dimethoxybenzyl alcohol, 2,4-dimethoxyphenyl methanol, benzenemethanol, 2,4-dimethoxy, 2,4-dimethoxybenzylalcohol, 1-hydroxymethyl-2,4-dimethoxybenzene, 2,4-dimethoxyphenyl methan-1-ol, pubchem7454, acmc-209opp, 2,4-dimethoxy-benzylalcohol, 2,4-dimethoxybenzyl-alcohol
(2,4-dimethoxyphenyl)methanol

Specifications

7314-44-5
100.0
>110°C
98.5% min. (GC)
C9H12O3
1.5450 to 1.5480
25g
2,4-dimethoxybenzyl alcohol, 2,4-dimethoxyphenyl methanol, benzenemethanol, 2,4-dimethoxy, 2,4-dimethoxybenzylalcohol, 1-hydroxymethyl-2,4-dimethoxybenzene, 2,4-dimethoxyphenyl methan-1-ol, pubchem7454, acmc-209opp, 2,4-dimethoxy-benzylalcohol, 2,4-dimethoxybenzyl-alcohol
COC1=CC(=C(C=C1)CO)OC
168.19
168.19
2, 4-Dimethoxybenzyl alcohol
98.5
177.0°C to 179.0°C (10.0mmHg)
Authentic
Glass bottle
(CH3O)2C6H3CH2OH
MFCD00004614
06, I, 550
RNKOUSCCPHSCFE-UHFFFAOYSA-N
(2,4-dimethoxyphenyl)methanol
81750
99%
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 230-775-2

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only