missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dimethoxybenzaldehyde 98.0+%, TCI America™

Catalog No. D062625G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D062625G 25 g
D0626100G 100 g
D0626500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D062625G Supplier TCI America Supplier No. D062625G

Chemical Identifiers

CAS 613-45-6
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.176
MDL Number MFCD00003311
InChI Key LWRSYTXEQUUTKW-UHFFFAOYSA-N
Synonym benzaldehyde, 2,4-dimethoxy, 2,4-dimethoxybenzaldehyd, 2,4-dimethoxy-benzaldehyde, 2,4-bis methyloxy benzaldehyde, ameba resin, pubchem8220, acmc-209mro, 2,4dimethoxybenzaldehyde, argopore tm-mb-cho, 2,4-dimethoxybenzaldehye
PubChem CID 69175
IUPAC Name 2,4-dimethoxybenzaldehyde
SMILES COC1=CC(=C(C=C1)C=O)OC

Specifications

Melting Point 71°C
Color White-Yellow
Boiling Point 165°C
Quantity 25 g
Formula Weight 166.18
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,4-Dimethoxybenzaldehyde
EINECSNumber (3)-2665
TSCA No