Learn More
2,4-Dimethoxybenzaldehyde, 98%
CAS: 613-45-6 | C9H10O3 | 166.176 g/mol
Supplier: Thermo Scientific Chemicals A1254936
Description
2,4-Dimethoxybenzaldehyde is used as a reagent to specifically quantify phlorotannins. It is also used as a component of hair dyes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2,4-Dimethoxybenzaldehyde is used as a reagent to specifically quantify phlorotannins. It is also used as a component of hair dyes.
Solubility
Insoluble in water. Soluble in chlorofrom.
Notes
Air sensitive. Incompatible with oxidizing agents.
Specifications
| 2,4-Dimethoxybenzaldehyde | |
| 67°C to 71°C | |
| C9H10O3 | |
| 500 g | |
| Air Sensitive | |
| Insoluble in water. Soluble in chlorofrom. | |
| COC1=CC(=C(C=C1)C=O)OC | |
| 166.176 | |
| 166.18 |
| 613-45-6 | |
| 165°C (10 mmHg) | |
| MFCD00003311 | |
| 607989 | |
| benzaldehyde, 2,4-dimethoxy, 2,4-dimethoxybenzaldehyd, 2,4-dimethoxy-benzaldehyde, 2,4-bis methyloxy benzaldehyde, ameba resin, pubchem8220, acmc-209mro, 2,4dimethoxybenzaldehyde, argopore tm-mb-cho, 2,4-dimethoxybenzaldehye | |
| LWRSYTXEQUUTKW-UHFFFAOYSA-N | |
| 2,4-dimethoxybenzaldehyde | |
| 69175 | |
| 98% |
Chemical Identifiers
| 613-45-6 | |
| 166.176 | |
| LWRSYTXEQUUTKW-UHFFFAOYSA-N | |
| 69175 | |
| COC1=CC(=C(C=C1)C=O)OC |
| C9H10O3 | |
| MFCD00003311 | |
| benzaldehyde, 2,4-dimethoxy, 2,4-dimethoxybenzaldehyd, 2,4-dimethoxy-benzaldehyde, 2,4-bis methyloxy benzaldehyde, ameba resin, pubchem8220, acmc-209mro, 2,4dimethoxybenzaldehyde, argopore tm-mb-cho, 2,4-dimethoxybenzaldehye | |
| 2,4-dimethoxybenzaldehyde |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 210-342-4
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only