missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dimethoxy-1,3,5-triazine 98.0+%, TCI America™

Catalog No. p-7128041
Change view
Click to view available options
Quantity:
1 g
5 g
Catalog No. Quantity
D39981G 1 g
D39985G 5 g
2 options

Catalog No. D39981G

Supplier: TCI America D39981G

Only null left

Chemical Identifiers

CAS 1898-72-2
Molecular Formula C5H7N3O2
Molecular Weight (g/mol) 141.13
InChI Key RIRBAVAYPRSMRH-UHFFFAOYSA-N
PubChem CID 243963
IUPAC Name 2,4-dimethoxy-1,3,5-triazine
SMILES COC1=NC(=NC=N1)OC

Specifications

Melting Point 59°C
Color White-Yellow
Quantity 1 g
Formula Weight 141.13
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,4-Dimethoxy-1,3,5-triazine