missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dihydroxypyridine 98.0+%, TCI America™

Catalog No. D26821G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D26821G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D26821G Supplier TCI America Supplier No. D26821G
Only null left

Chemical Identifiers

CAS 626-03-9
Molecular Formula C5H5NO2
Molecular Weight (g/mol) 111.1
MDL Number MFCD00006273
InChI Key ZEZJPIDPVXJEME-UHFFFAOYSA-N
Synonym 2,4-dihydroxypyridine, pyridine-2,4-diol, 2,4-pyridinediol, 4-hydroxypyridin-2 1h-one, 3-deazauracil, 4-hydroxy-2-pyridone, 2 1h-pyridinone, 4-hydroxy, 2,4-dihydroxy pyridine, 2-hydroxy-1h-pyridin-4-one, 4-hydroxy-2 1h-pyridinone
PubChem CID 54696004
IUPAC Name 4-hydroxy-1H-pyridin-2-one
SMILES C1=CNC(=O)C=C1O

Specifications

Color Yellow
Quantity 1 g
Formula Weight 111.10
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,4-Dihydroxypyridine
RTECSNumber UV1146800
TSCA No