missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dihydroxypyridine, 97%

Catalog No. AAA1356603
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1356603 1 g
AAA1356606 5 g
AAA1356614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1356603 Supplier Thermo Scientific Chemicals Supplier No. A1356603
Only null left

CAS: 626-03-9 | C5H5NO2 | 111.1 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 626-03-9
Molecular Formula C5H5NO2
Molecular Weight (g/mol) 111.1
MDL Number MFCD00006273
InChI Key ZEZJPIDPVXJEME-UHFFFAOYSA-N
Synonym 2,4-dihydroxypyridine, pyridine-2,4-diol, 2,4-pyridinediol, 4-hydroxypyridin-2 1h-one, 3-deazauracil, 4-hydroxy-2-pyridone, 2 1h-pyridinone, 4-hydroxy, 2,4-dihydroxy pyridine, 2-hydroxy-1h-pyridin-4-one, 4-hydroxy-2 1h-pyridinone
PubChem CID 54696004
IUPAC Name 4-hydroxy-1H-pyridin-2-one
SMILES C1=CNC(=O)C=C1O

Specifications

Melting Point ∼267°C (decomposition)
Quantity 1 g
Beilstein 108533
Formula Weight 111.1
Percent Purity 97%
Chemical Name or Material 2,4-Dihydroxypyridine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 210-924-8
RTECSNumber UV1146800
TSCA No

RUO – Research Use Only