missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(±)-2,4-Dihydroxybutyric acid lithium salt, ≥95.0% (GC), MilliporeSigma™ Supelco™
Specifications
Specifications
| CAS | 1518-62-3 |
| Linear Formula | C4H8O4 |
| Quantity | 10 mg |
| Molecular Weight (g/mol) | 120.10 |
| Formula Weight | 120.10 |
| Percent Purity | ≥95.0% (GC) |