missing translation for 'onlineSavingsMsg'
Learn More

2',4'-Dihydroxyacetophenone 98.0+%, TCI America™

Catalog No. D056125G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D056125G 25 g
D0561100G 100 g
D0561500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D056125G Supplier TCI America Supplier No. D056125G

Chemical Identifiers

CAS 89-84-9
Molecular Formula C8H8O3
Molecular Weight (g/mol) 152.15
MDL Number MFCD00002279
InChI Key SULYEHHGGXARJS-UHFFFAOYSA-N
Synonym 2',4'-dihydroxyacetophenone, 2,4-dihydroxyacetophenone, 1-2,4-dihydroxyphenyl ethanone, resacetophenone, 4-acetylresorcinol, resoacetophenone, ethanone, 1-2,4-dihydroxyphenyl, 1-acetyl-2,4-dihydroxybenzene, resorcinol, 4-acetyl, 1-2,4-dihydroxyphenyl ethan-1-one
PubChem CID 6990
ChEBI CHEBI:18414
IUPAC Name 1-(2,4-dihydroxyphenyl)ethan-1-one
SMILES CC(=O)C1=CC=C(O)C=C1O

Specifications

Melting Point 145°C
Quantity 25 g
Formula Weight 152.15
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2′,4′-Dihydroxyacetophenone
EINECSNumber (3)-1250
RTECSNumber AM7525000
TSCA Yes