missing translation for 'onlineSavingsMsg'
Learn More

2,4-Difluorobenzylzinc bromide, 0.5M in THF, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific™

Catalog No. aah58036ad
Change view
Click to view available options
Quantity:
50 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH58036AD 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH58036AD Supplier Thermo Scientific Chemicals Supplier No. H58036AD
Only null left

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
2,4-Difluorobenzylzinc bromide is used as pharmaceutical intermediates.

Solubility
Reacts with water.

Notes
Store at 4°C. Protect from heat. Store away from air. Air Sensitive. Do not store toge

Chemical Identifiers

CAS 307496-26-0
Molecular Formula C7H5BrF2Zn
Molecular Weight (g/mol) 272.398
MDL Number MFCD01311423
InChI Key WPJAHFYOACWPNA-UHFFFAOYSA-M
Synonym 2,4-difluorobenzylzinc bromide, bromo 2,4-difluorophenyl methyl zinc, 2,4-difluorobenzylzinc bromide 0.5m solution in thf, 2,4-difluorobenzylzinc bromide 0.5 m in tetrahydrofuran, 2,4-difluorobenzylzinc bromide solution, 0.5 m in thf, 2,4-difluorobenzylzinc bromide, 0.5m in thf, packaged under argon in resealable chemseal bottles
PubChem CID 2778322
IUPAC Name bromozinc(1+);2,4-difluoro-1-methanidylbenzene
SMILES [CH2-]C1=C(C=C(C=C1)F)F.[Zn+]Br

Specifications

Density 0.995
Quantity 50 mL
UN Number UN3399
Sensitivity Air sensitive; May form a precipitate
Solubility Information Reacts with water.
Formula Weight 272.4
Chemical Name or Material 2,4-Difluorobenzylzinc bromide, Packaged under Argon in resealable ChemSeal™ bottles
Hazard Category H225-H261-H302-H314-H335-H336-H351
Hazard Statement GHS H Statement
H225-H261-H314-H351-H335
Highly flammable liquid and vapor.
In contact with water releases flammable gas.
Causes severe skin burns and eye damage.
Suspected of causing cancer.
May cause respiratory irritation.
Precautionary Statement P201-P202-P210-P223-P231+P232-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P335+P334-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 4.3; Packing Group: II; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, LIQUID, WATER-REACTIVE, FLAMMABLE
TSCA No
Recommended Storage Keep cold; Store under Argon