missing translation for 'onlineSavingsMsg'
Learn More

2,4-Difluorobenzenesulfonyl Chloride 98.0+%, TCI America™

Catalog No. D29395G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D29395G 5 g
D293925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D29395G Supplier TCI America Supplier No. D29395G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 13918-92-8
Molecular Formula C6H3ClF2O2S
Molecular Weight (g/mol) 212.595
MDL Number MFCD00042481
InChI Key FJSAJUXIHJIAMD-UHFFFAOYSA-N
Synonym 2,4-difluorobenzene-1-sulfonyl chloride, 2,4-difluorobenzenesulphonyl chloride, 2,4-difluoro-benzenesulfonyl chloride, benzenesulfonyl chloride, 2,4-difluoro, 2,4-difluorophenyl chlorosulfone, buttpark 23\07-79, 2,4-difluorophenylsulfonyl chloride, pubchem5122, acmc-209civ
PubChem CID 2734273
IUPAC Name 2,4-difluorobenzenesulfonyl chloride
SMILES C1=CC(=C(C=C1F)F)S(=O)(=O)Cl

Specifications

Color Yellow
Boiling Point 84°C
Quantity 5 g
UN Number 3265
Formula Weight 212.59
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2,4-Difluorobenzenesulfonyl Chloride