missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2',4'-Difluoroacetanilide 98.0+%, TCI America™
Supplier: TCI America D357825G
Specifications
| 2′,4′-Difluoroacetanilide | |
| 122°C | |
| C8H7F2NO | |
| 25 g | |
| WOHLPEUHFSHZAN-UHFFFAOYSA-N | |
| N-(2,4-difluorophenyl)acetamide | |
| 96093 | |
| ≥98.0% (GC) |
| 399-36-0 | |
| Purple-White | |
| MFCD00032502 | |
| 2',4'-difluoroacetanilide, n-2,4-difluorophenyl acetamide, 2,4-difluoroacetanilide, 2,4-difluorophenyl acetamide, acetanilide,2,4-difluoro, acetamide, n-2,4-difluorophenyl, acetamide,n-2,4-difluorophenyl, aminobenzene, n-acetyl-2,4-difluoro, acetanilide, 2',4'-difluoro, maybridge1_001023 | |
| CC(=O)NC1=C(C=C(C=C1)F)F | |
| 171.147 | |
| 171.15 | |
| Crystalline Powder |
Chemical Identifiers
| 399-36-0 | |
| 171.147 | |
| WOHLPEUHFSHZAN-UHFFFAOYSA-N | |
| 96093 | |
| CC(=O)NC1=C(C=C(C=C1)F)F |
| C8H7F2NO | |
| MFCD00032502 | |
| 2',4'-difluoroacetanilide, n-2,4-difluorophenyl acetamide, 2,4-difluoroacetanilide, 2,4-difluorophenyl acetamide, acetanilide,2,4-difluoro, acetamide, n-2,4-difluorophenyl, acetamide,n-2,4-difluorophenyl, aminobenzene, n-acetyl-2,4-difluoro, acetanilide, 2',4'-difluoro, maybridge1_001023 | |
| N-(2,4-difluorophenyl)acetamide |
Safety and Handling
TSCA : No