Learn More
2,4-Difluoro-3-nitrobenzonitrile, 97%, Thermo Scientific™
Chemical Identifiers
| CAS | 1186194-75-1 |
|---|---|
| Molecular Formula | C7H2F2N2O2 |
| Molecular Weight (g/mol) | 184.1 |
| InChI Key | HESWWHRCQMLPFT-UHFFFAOYSA-N |
| Synonym | 3-nitro-2,4-difluoro-benzonitrile |
| PubChem CID | 45790497 |
| IUPAC Name | 2,4-difluoro-3-nitrobenzonitrile |
| SMILES | C1=CC(=C(C(=C1C#N)F)[N+](=O)[O-])F |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AC451420010
|
Thermo Scientific Chemicals
451420010 |
1g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 1186194-75-1 | |
| 184.1 | |
| 3-nitro-2,4-difluoro-benzonitrile | |
| 2,4-difluoro-3-nitrobenzonitrile |
| C7H2F2N2O2 | |
| HESWWHRCQMLPFT-UHFFFAOYSA-N | |
| 45790497 | |
| C1=CC(=C(C(=C1C#N)F)[N+](=O)[O-])F |
Specifications
| 1186194-75-1 | |
| Glass Bottle | |
| 1.529 | |
| 3-nitro-2,4-difluoro-benzonitrile | |
| C1=CC(=C(C(=C1C#N)F)[N+](=O)[O-])F | |
| 184.1 | |
| 184.1 | |
| 2, 4-Difluoro-3-nitrobenzonitrile |
| 96% min. (GC) | |
| C7H2F2N2O2 | |
| 1g | |
| HESWWHRCQMLPFT-UHFFFAOYSA-N | |
| 2,4-difluoro-3-nitrobenzonitrile | |
| 45790497 | |
| 97% |
Safety and Handling
GHS H Statement
Toxic if swallowed.
Toxic in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Harmful if inhaled.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of
GHS Signal Word: Danger
RUO â Research Use Only