missing translation for 'onlineSavingsMsg'
Learn More

2',4'-Difluoro-2-(1H-1,2,4-triazol-1-yl)acetophenone, 97%

Catalog No. p-7054032
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6188706 5 g
AAH6188714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6188706 Supplier Thermo Scientific Chemicals Supplier No. H6188706

CAS: 86404-63-9 | C10H7F2N3O | 223.183 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 86404-63-9
Molecular Formula C10H7F2N3O
Molecular Weight (g/mol) 223.183
MDL Number MFCD02093825
InChI Key XCHRPVARHBCFMJ-UHFFFAOYSA-N
Synonym 1-2,4-difluorophenyl-2-1h-1,2,4-triazol-1-yl ethanone, 2,4-difluoro-alpha-1h-1,2,4-triazolyl acetophenone, 1-2,4-difluorophenyl-2-1,2,4-triazol-1-yl ethanone, 2',4'-difluoro-2-1h-1,2,4-triazol-1-yl acetophenone, unii-hxi8r9r915, 2-1h-1,2,4-triazol-1-yl-2',4'-difluoroacetophenone, ethanone, 1-2,4-difluorophenyl-2-1h-1,2,4-triazol-1-yl, 2',4'-difluoro-2-1h-1,2,4-triazolyl acetophenone, 1-2,4-difluorophenyl-2-1h-1,2,4-triazol-1-yl ethan-1-one, 1-2,4-difluorophenyl-2-1h-1,2,4-triazol-1-yl-ethanone
PubChem CID 588080
IUPAC Name 1-(2,4-difluorophenyl)-2-(1,2,4-triazol-1-yl)ethanone
SMILES C1=CC(=C(C=C1F)F)C(=O)CN2C=NC=N2

Specifications

Melting Point 103°C to 107°C
Color Brown to White
Quantity 5 g
Formula Weight 223.18
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 2',4'-Difluoro-2-(1H-1,2,4-triazol-1-yl)acetophenone
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only