missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichlorophenylboronic acid, 97%, Thermo Scientific™

Chemical Identifiers

CAS 68716-47-2
Molecular Formula C6H5BCl2O2
Molecular Weight (g/mol) 190.81
MDL Number MFCD00013930
InChI Key QNEGDGPAXKYZHZ-UHFFFAOYSA-N
Synonym 2,4-dichlorophenyl boronic acid, 2,4-dichlorobenzeneboronic acid, 2,4-dichlorophenylbornic acid, 2,4-dichlorophenyl boranediol, chembl20726, 2,4-dichloro benzene boronic acid, boronic acid, 2,4-dichlorophenyl, 2,4-dichlorobenzeneboron dihydroxide, 2,4-dichlorobenzeneboronicacid
PubChem CID 2734659
IUPAC Name (2,4-dichlorophenyl)boronic acid
SMILES OB(O)C1=CC=C(Cl)C=C1Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC367670010
SDS View Documents
Thermo Scientific Chemicals
367670010
1g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC367670050
SDS View Documents
Thermo Scientific Chemicals
367670050
5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

68716-47-2
190.81
QNEGDGPAXKYZHZ-UHFFFAOYSA-N
2734659
OB(O)C1=CC=C(Cl)C=C1Cl
C6H5BCl2O2
MFCD00013930
2,4-dichlorophenyl boronic acid, 2,4-dichlorobenzeneboronic acid, 2,4-dichlorophenylbornic acid, 2,4-dichlorophenyl boranediol, chembl20726, 2,4-dichloro benzene boronic acid, boronic acid, 2,4-dichlorophenyl, 2,4-dichlorobenzeneboron dihydroxide, 2,4-dichlorobenzeneboronicacid
(2,4-dichlorophenyl)boronic acid

Specifications

68716-47-2
100.0
97%
C6H5BCl2O2
1g
Solubility in water: insoluble. Other solubilities: soluble in methanol
OB(O)C1=CC=C(Cl)C=C1Cl
190.81
190.82
2, 4-Dichlorophenylboronic acid, 97%
96.0
Authentic
Glass bottle
MFCD00013930
2,4-dichlorophenyl boronic acid, 2,4-dichlorobenzeneboronic acid, 2,4-dichlorophenylbornic acid, 2,4-dichlorophenyl boranediol, chembl20726, 2,4-dichloro benzene boronic acid, boronic acid, 2,4-dichlorophenyl, 2,4-dichlorobenzeneboron dihydroxide, 2,4-dichlorobenzeneboronicacid
QNEGDGPAXKYZHZ-UHFFFAOYSA-N
(2,4-dichlorophenyl)boronic acid
2734659
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face prot

GHS Signal Word: Warning

Recommended Storage : Refrigerator +4°C

SDS
Documents

Documents

RUO – Research Use Only