missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichlorophenylacetic Acid 98.0+%, TCI America™

Catalog No. D181225G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D181225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D181225G Supplier TCI America Supplier No. D181225G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 19719-28-9
Molecular Formula C8H6Cl2O2
Molecular Weight (g/mol) 205.034
MDL Number MFCD00004318
InChI Key GXMWLJKTGBZMBH-UHFFFAOYSA-N
Synonym 2,4-dichlorophenylacetic acid, 2-2,4-dichlorophenyl acetic acid, benzeneacetic acid, 2,4-dichloro, 2,4-dichloro-phenyl-acetic acid, 2,4-dichlorophenyl acetic acid, 2 ,4-dichlorophenylacetic acid, zlchem 477, pubchem15026, acmc-209f0x
PubChem CID 88209
IUPAC Name 2-(2,4-dichlorophenyl)acetic acid
SMILES C1=CC(=C(C=C1Cl)Cl)CC(=O)O

Specifications

Melting Point 132°C
Color White-Yellow
Quantity 25 g
Formula Weight 205.03
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2,4-Dichlorophenylacetic Acid
EINECSNumber (3)-3780
TSCA No