missing translation for 'onlineSavingsMsg'
Learn More

2',4'-Dichlorobutyrophenone 96.0+%, TCI America™

Catalog No. D346825G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D346825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D346825G Supplier TCI America Supplier No. D346825G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 66353-47-7
Molecular Formula C10H10Cl2O
Molecular Weight (g/mol) 217.089
InChI Key FXCJYMLJTUZGDU-UHFFFAOYSA-N
Synonym 2,4-Dichlorophenyl Propyl Ketone
PubChem CID 13175617
IUPAC Name 1-(2,4-dichlorophenyl)butan-1-one
SMILES CCCC(=O)C1=C(C=C(C=C1)Cl)Cl

Specifications

Boiling Point 141°C
Quantity 25 g
Formula Weight 217.09
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 2′,4′-Dichlorobutyrophenone