missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichlorobenzylmagnesium chloride, 0.25M in 2-MeTHF

Catalog No. AAH54159AE
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH54159AE 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH54159AE Supplier Thermo Scientific Chemicals Supplier No. H54159AE
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 129752-86-9 | C7H5Cl3Mg | 219.77 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 129752-86-9
Molecular Formula C7H5Cl3Mg
Molecular Weight (g/mol) 219.77
MDL Number MFCD01319913
InChI Key MJNFFJCHKWQLCN-UHFFFAOYSA-M
Synonym 2,4-dichlorobenzylmagnesium chloride, 2,4-dichlorobenzylmagnesium chloride, 0.25m in 2-methf, chloro 2,4-dichlorophenyl methyl magnesium, mjnffjchkwqlcn-uhfffaoysa-m, 2,4-dichlorobenzyl magnesium chloride, 2,4-dichlorobenzyl-magnesium chloride, magnesium,2,4-dichloro-1-methanidylbenzene,chloride, 2,4-dichlorobenzylmagnesium chloride 0.25 m in diethyl ether
PubChem CID 13120922
IUPAC Name magnesium;2,4-dichloro-1-methanidylbenzene;chloride
SMILES Cl[Mg]CC1=C(Cl)C=C(Cl)C=C1

Specifications

Quantity 100 mL
UN Number UN2924
Sensitivity Air and moisture sensitive; May form a precipitate
Formula Weight 219.78
Concentration or Composition (by Analyte or Components) 0.25M in 2-MeTHF
Chemical Name or Material 2,4-Dichlorobenzylmagnesium chloride
Hazard Category H225-H302-H314-H335
Hazard Statement GHS H Statement
H225-H314
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
TSCA No
Recommended Storage Keep cold; Store under Argon

RUO – Research Use Only