missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichlorobenzyl Mercaptan 98.0+%, TCI America™

Catalog No. p-7127165
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D21985G 5 g
D219825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D21985G Supplier TCI America Supplier No. D21985G
Only null left

Chemical Identifiers

CAS 59293-67-3
Molecular Formula C7H6Cl2S
Molecular Weight (g/mol) 193.085
MDL Number MFCD00039650
InChI Key ZSPXTTVUJDSRNJ-UHFFFAOYSA-N
Synonym 2,4-dichlorobenzyl mercaptan, 2,4-dichlorophenyl methanethiol, 2,4-dichlorobenzylmercaptan, 2,4-dichlorobenzenemethanethiol, 2,4-dichlorotoluene-alpha-thiol, benzenemethanethiol, 2,4-dichloro, 2,4-dichloro-alpha-toluenethiol, acmc-1b9kz, d0sz9f, 2,4-dichlorothiobenzyl alcohol
PubChem CID 95756
IUPAC Name (2,4-dichlorophenyl)methanethiol
SMILES C1=CC(=C(C=C1Cl)Cl)CS

Specifications

Color Yellow
Boiling Point 115°C
Quantity 5 g
UN Number 2810
Formula Weight 193.09
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,4-Dichlorobenzyl Mercaptan