missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichlorobenzonitrile 98.0+%, TCI America™

Catalog No. D33495G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D33495G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D33495G Supplier TCI America Supplier No. D33495G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6574-98-7
Molecular Formula C7H3Cl2N
Molecular Weight (g/mol) 172.008
InChI Key GRUHREVRSOOQJG-UHFFFAOYSA-N
Synonym benzonitrile, 2,4-dichloro, 2,4-dichloro-benzonitrile, 2,4-dichlorobenzenecarbonitrile, acmc-209nrx, 2,4-dichloro benzonitrile, ksc352s3f
PubChem CID 81050
IUPAC Name 2,4-dichlorobenzonitrile
SMILES C1=CC(=C(C=C1Cl)Cl)C#N

Specifications

Melting Point 61°C
Color White
Boiling Point 125°C
Quantity 5 g
UN Number 3439
Formula Weight 172.01
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,4-Dichlorobenzonitrile