missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichlorobenzeneboronic acid, 98%

Catalog No. p-7043607
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0156303 1 g
AAL0156306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0156303 Supplier Thermo Scientific Chemicals Supplier No. L0156303
Only null left

CAS: 68716-47-2 | C6H5BCl2O2 | 190.81 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 68716-47-2
Molecular Formula C6H5BCl2O2
Molecular Weight (g/mol) 190.81
MDL Number MFCD00013930
InChI Key QNEGDGPAXKYZHZ-UHFFFAOYSA-N
Synonym 2,4-dichlorophenyl boronic acid, 2,4-dichlorobenzeneboronic acid, 2,4-dichlorophenylbornic acid, 2,4-dichlorophenyl boranediol, chembl20726, 2,4-dichloro benzene boronic acid, boronic acid, 2,4-dichlorophenyl, 2,4-dichlorobenzeneboron dihydroxide, 2,4-dichlorobenzeneboronicacid
PubChem CID 2734659
IUPAC Name (2,4-dichlorophenyl)boronic acid
SMILES OB(O)C1=CC=C(Cl)C=C1Cl

Specifications

Melting Point 242°C to 245°C
Quantity 1 g
Beilstein 4983878
Formula Weight 190.82
Percent Purity ≥98%
Chemical Name or Material 2,4-Dichlorobenzeneboronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only