missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichlorobenzaldehyde 95.0+%, TCI America™

Catalog No. d033025g
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
500 g
Catalog No. Quantity
D033025G 25 g
D0330500G 500 g
2 options

Catalog No. D033025G

Supplier: TCI America D033025G

Only null left

Chemical Identifiers

CAS 874-42-0
Molecular Formula C7H4Cl2O
Molecular Weight (g/mol) 175.008
MDL Number MFCD00003305
InChI Key YSFBEAASFUWWHU-UHFFFAOYSA-N
Synonym benzaldehyde, 2,4-dichloro, unii-z08ql2124r, ccris 6013, 2,4-dichloro benzaldehyde, 2,4-dichloro-benzaldehyde, acmc-209qnb, dsstox_cid_2130, 2,4 dichloro benzaldehyde, 2,4-di-chlorobenzaldehyde, dsstox_rid_76500
PubChem CID 13404
IUPAC Name 2,4-dichlorobenzaldehyde
SMILES C1=CC(=C(C=C1Cl)Cl)C=O

Specifications

Melting Point 72°C
Color White-Yellow
Boiling Point 233°C
Quantity 25 g
UN Number 1759
Formula Weight 175.01
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,4-Dichlorobenzaldehyde
EINECSNumber (3)-3636
TSCA Yes