missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichloro-L-phenylalanine, 98%, Thermo Scientific™

Catalog No. p-7030048
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2757903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH2757903 Supplier Thermo Scientific Chemicals Supplier No. H2757903

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 111119-36-9
Molecular Formula C9H9Cl2NO2
Molecular Weight (g/mol) 234.08
MDL Number MFCD01860882
InChI Key GWHQTNKPTXDNRM-QMMMGPOBSA-N
Synonym 2,4-dichloro-l-phenylalanine, l-2,4-dichlorophenylalanine, 2s-2-amino-3-2,4-dichlorophenyl propanoic acid, s-2-amino-3-2,4-dichlorophenyl propanoic acid, h-phe 2,4-dicl-oh, l-2,4-dichlorophe, l-phenylalanine, 2,4-dichloro, h-phe 2,4-cl2-oh, 2,4-dichloro-phenylalanine, s-2-amino-3-2,4-dichloro-phenyl-propionic acid
PubChem CID 2761497
IUPAC Name (2S)-2-amino-3-(2,4-dichlorophenyl)propanoic acid
SMILES N[C@@H](CC1=CC=C(Cl)C=C1Cl)C(O)=O

Specifications

Quantity 1 g
Formula Weight 234.09
Percent Purity 98%
Chemical Name or Material 2,4-Dichloro-L-phenylalanine
TSCA No
Recommended Storage Ambient temperatures