missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4-Dichloro-6-phenyl-1,3,5-triazine 98.0+%, TCI America™
$33.56 - $100.67
Chemical Identifiers
| CAS | 1700-02-3 |
|---|---|
| Molecular Formula | C9H5Cl2N3 |
| Molecular Weight (g/mol) | 226.06 |
| MDL Number | MFCD00047340 |
| InChI Key | AMEVJOWOWQPPJQ-UHFFFAOYSA-N |
| PubChem CID | 15549 |
| IUPAC Name | 2,4-dichloro-6-phenyl-1,3,5-triazine |
| SMILES | C1=CC=C(C=C1)C2=NC(=NC(=N2)Cl)Cl |
Chemical Identifiers
| 1700-02-3 | |
| 226.06 | |
| AMEVJOWOWQPPJQ-UHFFFAOYSA-N | |
| 2,4-dichloro-6-phenyl-1,3,5-triazine |
| C9H5Cl2N3 | |
| MFCD00047340 | |
| 15549 | |
| C1=CC=C(C=C1)C2=NC(=NC(=N2)Cl)Cl |
Specifications
| 1700-02-3 | |
| White-Yellow | |
| C9H5Cl2N3 | |
| 1 g | |
| C1=CC=C(C=C1)C2=NC(=NC(=N2)Cl)Cl | |
| 226.06 | |
| 226.06 | |
| Crystalline Powder |
| 121°C | |
| 136°C | |
| MFCD00047340 | |
| AMEVJOWOWQPPJQ-UHFFFAOYSA-N | |
| 2,4-dichloro-6-phenyl-1,3,5-triazine | |
| 15549 | |
| ≥98.0% (GC) | |
| 2,4-Dichloro-6-phenyl-1,3,5-triazine |
Safety and Handling
TSCA : No