missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichloro-6-methylpyrimidine 98.0+%, TCI America™

Catalog No. D29585G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D29585G 5 g
D295825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D29585G Supplier TCI America Supplier No. D29585G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5424-21-5
Molecular Formula C5H4Cl2N2
Molecular Weight (g/mol) 163.001
MDL Number MFCD00006064
InChI Key BTLKROSJMNFSQZ-UHFFFAOYSA-N
Synonym pyrimidine, 2,4-dichloro-6-methyl, 2,6-dichloro-4-methylpyrimidine, 2,4-dichloro-6-methyl-pyrimidine, 2,4-dichloro-6-methyl pyrimidine, 6-methyl-2,4-dichloropyrimidine, 2,4-dichlor-6-methylpyrimidin, pubchem6911, acmc-1b11m, ksc495i8r, 2,4-dichloro6-methylpyrimidine
PubChem CID 79471
IUPAC Name 2,4-dichloro-6-methylpyrimidine
SMILES CC1=CC(=NC(=N1)Cl)Cl

Specifications

Melting Point 47°C
Color White-Yellow
Boiling Point 219°C
Quantity 5 g
Formula Weight 163.00
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,4-Dichloro-6-methylpyrimidine